The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194357.1 | NP_919338.2 | 14 - 15 | 1452 | 90 |
GenBank mRNA | AK132627.1 | - | 9 - 10 | 804 | 90 |
BC048084.1 | - | 14 - 15 | 1452 | 90 |