The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019758.2 | NP_062732.1 | 7 - 8 | 621 | 161 |
GenBank mRNA | AF176009.1 | - | 7 - 8 | 519 | 161 |
AK003100.1 | - | 7 - 8 | 488 | 161 | |
AK007757.1 | - | 7 - 8 | 491 | 161 | |
AK008463.1 | - | 7 - 8 | 490 | 161 | |
AK008487.1 | - | 7 - 8 | 495 | 161 | |
AK008712.1 | - | 7 - 8 | 622 | 161 | |
AK011785.1 | - | 7 - 8 | 490 | 161 | |
AK011814.1 | - | 7 - 8 | 573 | 161 | |
AK011874.1 | - | 7 - 8 | 498 | 161 | |
AK075890.1 | - | 7 - 8 | 490 | 161 | |
AK151553.1 | - | 7 - 8 | 499 | 161 | |
AK161441.1 | - | 7 - 8 | 494 | 161 | |
AK164169.1 | - | 7 - 8 | 506 | 161 | |
BC002152.1 | - | 5 - 6 | 306 | 161 | |
BC028929.1 | - | 6 - 7 | 513 | 161 | |
BC038899.1 | - | 7 - 8 | 528 | 161 | |
BC038916.1 | - | 7 - 8 | 528 | 161 | |
BC065145.1 | - | 7 - 8 | 507 | 161 |