The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025597.2 | NP_079873.1 | 2 - 3 | 229 | 157 |
GenBank mRNA | AK007930.1 | - | 2 - 3 | 228 | 157 |
AK009008.1 | - | 2 - 3 | 191 | 157 | |
AK012320.1 | - | 2 - 3 | 230 | 157 | |
AK013608.1 | - | 2 - 3 | 215 | 157 | |
AK138123.1 | - | 2 - 3 | 227 | 157 | |
BC028669.1 | - | 2 - 3 | 385 | 157 | |
BC036989.1 | - | 2 - 3 | 226 | 157 |