Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ndufa6
- Entrez Gene ID
-
67130
- Gene Name
- NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 6 (B14)
- Gene Aliases
-
14kDa, 2700038D15Rik, B230217P19Rik
- Chromosome Location
-
Chr.15: 82350139 - 82354291 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BC038336.1 |
- |
|
|
|
Chr. - - [-] on Build |
133 |
View Full Product Details