Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmn1r56
Entrez Gene ID
81015
Gene Name
vomeronasal 1 receptor 56
Gene Aliases
V1rd3, V3R3
Chromosome Location
Chr.7: 5194916 - 5196747 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030740.1 NP_109665.1 1 - 1 469 246
GenBank mRNA AF324868.1 - 1 - 1 469 246

View Full Product Details