The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153388.4 | NP_700437.2 | 2 - 3 | 2060 | 201 |
GenBank mRNA | AK081560.1 | - | 1 - 2 | 2343 | 201 |
AK163603.1 | - | 2 - 3 | 1918 | 201 | |
BC023036.1 | - | 1 - 2 | 1673 | 201 | |
BC023156.1 | - | 2 - 3 | 1716 | 201 | |
DQ078786.1 | - | 1 - 2 | 1345 | 201 |