The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276315.1 | NP_001263244.1 | 10 - 11 | 1953 | 97 |
NM_016912.2 | NP_058608.1 | 10 - 11 | 1687 | 97 | |
NM_177270.4 | NP_796244.2 | 10 - 11 | 1687 | 97 | |
GenBank mRNA | AB029065.1 | - | 10 - 11 | 1663 | 97 |
AB029066.1 | - | 10 - 11 | 1651 | 97 | |
AK030598.1 | - | 10 - 11 | 1942 | 97 | |
AK139820.1 | - | 10 - 11 | 1896 | 97 | |
AK144574.1 | - | 10 - 11 | 1688 | 97 | |
BC117894.1 | - | 10 - 11 | 1596 | 97 | |
BC117895.1 | - | 10 - 11 | 1596 | 97 |