The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009960.1 | NP_034090.1 | 1 - 2 | 2395 | 71 |
GenBank mRNA | AB008182.1 | - | 1 - 2 | 2288 | 71 |
AK043705.1 | - | 1 - 2 | 71 | ||
AK139436.1 | - | 1 - 2 | 2541 | 71 | |
AY013758.1 | - | 1 - 2 | 2395 | 71 | |
BC141349.1 | - | 1 - 2 | 2421 | 71 |