The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172943.4 | NP_766531.2 | 2 - 3 | 1586 | 74 |
GenBank mRNA | AK049130.1 | - | 2 - 3 | 1279 | 74 |
AK053695.1 | - | 2 - 3 | 1292 | 74 | |
AK053700.1 | - | 2 - 3 | 1159 | 74 | |
AK162072.1 | - | 2 - 3 | 1160 | 74 | |
AK163294.1 | - | 2 - 3 | 1159 | 74 | |
AK169700.1 | - | 2 - 3 | 1221 | 74 | |
BC052076.1 | - | 2 - 3 | 1193 | 74 |