The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178932.1 | NP_849263.1 | 1 - 2 | 1614 | 74 |
GenBank mRNA | AF350445.2 | - | 1 - 2 | 1614 | 74 |
BC150843.1 | - | 1 - 2 | 1605 | 74 | |
BC150848.1 | - | 1 - 2 | 1605 | 74 |