The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012020.2 | NP_036150.1 | 1 - 1 | 1139 | 71 |
GenBank mRNA | BC137811.1 | - | 1 - 1 | 1139 | 71 |
BC137812.1 | - | 1 - 1 | 1139 | 71 |