The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010772.1 | NP_034902.1 | 2 - 3 | 1105 | 58 |
GenBank mRNA | BC004758.1 | - | 4 - 5 | 642 | 58 |
DQ150586.1 | - | 1 - 2 | 819 | 58 | |
L04649.1 | - | 2 - 3 | 1078 | 58 |