Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Mbnl2
- Entrez Gene ID
-
105559
- Gene Name
- muscleblind-like 2
- Gene Aliases
-
1110002M11Rik, AI047808, AI837313, AI849185, AL118326, MBLL, R75232, mKIAA4072
- Chromosome Location
-
Chr.14: 120275661 - 120431698 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC079898.1 |
- |
8 - 9 |
1681 |
78 |
View Full Product Details