Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dcdc2a
Entrez Gene ID
195208
Gene Name
doublecortin domain containing 2a
Gene Aliases
AW492955, Dcdc2, RU2
Chromosome Location
Chr.13: 25020435 - 25210706 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177577.3 NP_808245.2 8 - 9 1287 89

View Full Product Details