The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081267.2 | NP_001074736.1 | 4 - 5 | 599 | 120 |
GenBank mRNA | AK032853.1 | - | 4 - 5 | 575 | 120 |
AK146154.1 | - | 4 - 5 | 603 | 120 | |
AK146675.1 | - | 4 - 5 | 609 | 120 | |
BC085127.1 | - | 4 - 5 | 600 | 120 |