The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042767.2 | NP_001036232.1 | 5 - 6 | 480 | 62 |
NM_001042768.2 | NP_001036233.2 | 6 - 7 | 560 | 62 | |
NM_008934.3 | NP_032960.3 | 5 - 6 | 492 | 62 | |
NR_045173.1 | 5 - 6 | 496 | 62 | ||
GenBank mRNA | AF318182.1 | - | 5 - 6 | 469 | 62 |
BC013896.1 | - | 5 - 6 | 497 | 62 | |
CA468822.1 | - | 5 - 6 | 504 | 62 | |
CB599669.1 | - | 5 - 6 | 483 | 62 | |
CF550816.1 | - | 1 - 2 | 56 | 62 | |
D10445.1 | - | 4 - 5 | 408 | 62 | |
DV662175.1 | - | 4 - 5 | 394 | 62 |