The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001162941.1 | NP_001156413.1 | 7 - 8 | 988 | 71 |
NM_001162942.1 | NP_001156414.1 | 7 - 8 | 931 | 71 | |
NM_153058.4 | NP_694698.3 | 6 - 7 | 1039 | 71 | |
GenBank mRNA | AK028823.1 | - | 7 - 8 | 989 | 71 |
AK038562.1 | - | 6 - 7 | 1026 | 71 | |
AK040767.1 | - | 6 - 7 | 1014 | 71 | |
AK045493.1 | - | 6 - 7 | 922 | 71 | |
AK045904.1 | - | 6 - 7 | 1014 | 71 | |
AK046095.1 | - | 6 - 7 | 982 | 71 | |
AK046221.1 | - | 6 - 7 | 1006 | 71 | |
AK148954.1 | - | 6 - 7 | 1014 | 71 | |
BC025804.1 | - | 6 - 7 | 986 | 71 | |
BC027056.1 | - | 6 - 7 | 1005 | 71 | |
BC028987.1 | - | 6 - 7 | 961 | 71 | |
BC035254.1 | - | 6 - 7 | 991 | 71 |