Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rab5b
Entrez Gene ID
19344
Gene Name
RAB5B, member RAS oncogene family
Gene Aliases
C030027M18Rik
Chromosome Location
Chr.10: 128677172 - 128696306 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177411.4 NP_803130.1 1 - 2 77 79
GenBank mRNA AK081251.1 - 1 - 2 73 79
AK083034.1 - 1 - 2 66 79
BY727964.1 - 1 - 2 78 79

View Full Product Details