Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prkag2
Entrez Gene ID
108099
Gene Name
protein kinase, AMP-activated, gamma 2 non-catalytic subunit
Gene Aliases
2410051C13Rik, AAKG, AAKG2, AI854673, H91620p, WPWS
Chromosome Location
Chr.5: 24862735 - 25101906 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170555.1 NP_001164026.1 6 - 7 541 107
NM_001170556.1 NP_001164027.1 6 - 7 538 107
NM_145401.2 NP_663376.2 10 - 11 1620 107
GenBank mRNA AK032238.1 - 8 - 9 1132 107
AK034765.1 - 4 - 5 206 107
AK154296.1 - 10 - 11 1621 107
AK165834.1 - 6 - 7 480 107
AY348864.1 - 6 - 7 471 107
BC015283.1 - 10 - 11 1408 107
BQ572978.1 - 6 - 7 471 107
BU707674.1 - 6 - 7 441 107

View Full Product Details