Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prkag2
Entrez Gene ID
108099
Gene Name
protein kinase, AMP-activated, gamma 2 non-catalytic subunit
Gene Aliases
2410051C13Rik, AAKG, AAKG2, AI854673, H91620p, WPWS
Chromosome Location
Chr.5: 24862735 - 25101906 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170555.1 NP_001164026.1 9 - 10 867 100
NM_001170556.1 NP_001164027.1 9 - 10 864 100
NM_145401.2 NP_663376.2 13 - 14 1946 100
GenBank mRNA AK032238.1 - 11 - 12 1458 100
AK034765.1 - 7 - 8 532 100
AK154296.1 - 13 - 14 1947 100
AK165834.1 - 9 - 10 806 100
AY348864.1 - 9 - 10 797 100
BC015283.1 - 13 - 14 1734 100

View Full Product Details