Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prkag2
Entrez Gene ID
108099
Gene Name
protein kinase, AMP-activated, gamma 2 non-catalytic subunit
Gene Aliases
2410051C13Rik, AAKG, AAKG2, AI854673, H91620p, WPWS
Chromosome Location
Chr.5: 24862735 - 25101906 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170555.1 NP_001164026.1 5 - 6 477 148
NM_001170556.1 NP_001164027.1 5 - 6 474 148
NM_145401.2 NP_663376.2 9 - 10 1556 148
GenBank mRNA AK032238.1 - 7 - 8 1068 148
AK034765.1 - 3 - 4 142 148
AK154296.1 - 9 - 10 1557 148
AK165834.1 - 5 - 6 416 148
AY348864.1 - 5 - 6 407 148
BC015283.1 - 9 - 10 1344 148
BQ572978.1 - 5 - 6 407 148
BU707674.1 - 5 - 6 377 148

View Full Product Details