Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cd36
Entrez Gene ID
12491
Gene Name
CD36 antigen
Gene Aliases
FAT, GPIV, Scarb3
Chromosome Location
Chr.5: 17781690 - 17888959 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159555.1 NP_001153027.1 10 - 11 1468 71
NM_001159556.1 NP_001153028.1 10 - 11 1328 71
NM_001159557.1 NP_001153029.1 11 - 12 1379 71
NM_001159558.1 NP_001153030.1 11 - 12 1495 71
NM_007643.4 NP_031669.3 10 - 11 1471 71
GenBank mRNA AK052825.1 - 10 - 11 1272 71
AK128908.1 - 11 - 12 1272 71
AK150648.1 - 10 - 11 1292 71
AK150865.1 - 6 - 7 666 71
AK151355.1 - 10 - 11 1307 71
AK152642.1 - 5 - 6 463 71
AK152904.1 - 8 - 9 978 71
AK153095.1 - 5 - 6 581 71
AK172162.1 - 11 - 12 1317 71
BC010262.1 - 10 - 11 1280 71
HQ171908.1 - 1 - 2 42 71
L23108.1 - 10 - 11 1289 71

View Full Product Details