Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam3c
Entrez Gene ID
27999
Gene Name
family with sequence similarity 3, member C
Gene Aliases
D6Wsu176e, Ilei
Chromosome Location
Chr.6: 22306520 - 22356081 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138587.4 NP_613053.3 7 - 8 513 129
GenBank mRNA AK154498.1 - 7 - 8 543 129
AK170887.1 - 7 - 8 562 129
AY424275.1 - 7 - 8 513 129
BC009086.1 - 7 - 8 513 129
BC069880.1 - 6 - 7 639 129

View Full Product Details