The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145387.4 | NP_663362.2 | 13 - 14 | 2109 | 99 |
GenBank mRNA | AK077662.1 | - | 13 - 14 | 2110 | 99 |
AK085600.1 | - | 12 - 13 | 2280 | 99 | |
AK171694.1 | - | 11 - 12 | 1885 | 99 | |
BC056953.1 | - | 13 - 14 | 2071 | 99 |