Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankhd1
Entrez Gene ID
108857
Gene Name
ankyrin repeat and KH domain containing 1
Gene Aliases
1110004O12Rik, 4933432B13Rik, 9130019P20Rik, A530027J04Rik, A630021B20Rik, AA571404, Mask, mFLJ00246, mKIAA1085
Chromosome Location
Chr.18: 36560603 - 36658908 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175375.3 NP_780584.2 12 - 13 2148 66
GenBank mRNA AK017018.1 - 3 - 4 246 66
AK029322.1 - 5 - 6 563 66
AK131166.1 - 12 - 13 2011 66

View Full Product Details