Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxl13
Entrez Gene ID
320118
Gene Name
F-box and leucine-rich repeat protein 13
Gene Aliases
4921539K22Rik
Chromosome Location
Chr.5: 21483847 - 21645698 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199632.1 NP_001186561.1 15 - 16 1705 150
NM_177076.4 NP_796050.2 14 - 15 1606 150
GenBank mRNA AK029566.1 - 14 - 15 1608 150
AK030172.1 - 15 - 16 1688 150
BC145291.1 - 15 - 16 1631 150
BC145292.1 - 14 - 15 1533 150

View Full Product Details