Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxl13
Entrez Gene ID
320118
Gene Name
F-box and leucine-rich repeat protein 13
Gene Aliases
4921539K22Rik
Chromosome Location
Chr.5: 21483847 - 21645698 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199632.1 NP_001186561.1 16 - 17 1830 90
NM_177076.4 NP_796050.2 15 - 16 1731 90
GenBank mRNA AK029566.1 - 15 - 16 1733 90
BC145291.1 - 16 - 17 1756 90
BC145292.1 - 15 - 16 1658 90

View Full Product Details