Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vps11
Entrez Gene ID
71732
Gene Name
vacuolar protein sorting 11 (yeast)
Gene Aliases
1200011A11Rik
Chromosome Location
Chr.9: 44347881 - 44361675 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027889.1 NP_082165.1 10 - 11 1798 109
GenBank mRNA AK004695.1 - 10 - 11 1773 109
AK032484.1 - 10 - 11 1763 109
AK090030.1 - 10 - 11 1779 109
AK153963.1 - 10 - 11 1771 109
AK162897.1 - 10 - 11 1773 109
BC029004.1 - 10 - 11 1798 109

View Full Product Details