The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003949.3 | NP_001003949.2 | 10 - 11 | 1488 | 94 |
GenBank mRNA | AK131170.1 | - | - | 205 | 94 |
AY096036.1 | - | 10 - 11 | 1198 | 94 | |
AY096037.1 | - | 9 - 10 | 1270 | 94 | |
BC005494.1 | - | 9 - 10 | 1011 | 94 | |
BC052787.1 | - | 9 - 10 | 1030 | 94 | |
BC056654.1 | - | 1 - 2 | 341 | 94 | |
BC080302.1 | - | - | 691 | 94 | |
BC100297.1 | - | 7 - 8 | 625 | 94 |