Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Npsr1
Entrez Gene ID
319239
Gene Name
neuropeptide S receptor 1
Gene Aliases
9330128H10Rik, GPRA, Gpr154, MVTR, PGR14, VRR1
Chromosome Location
Chr.9: 24097950 - 24317590 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175678.3 NP_783609.1 5 - 6 573 71
GenBank mRNA AK033957.1 - 5 - 6 574 71
AK041451.1 - 4 - 5 472 71
AY299387.1 - 4 - 5 509 71

View Full Product Details