Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankrd24
Entrez Gene ID
70615
Gene Name
ankyrin repeat domain 24
Gene Aliases
4631433D01Rik, 5730519E19Rik, D10Bur2e
Chromosome Location
Chr.10: 81627795 - 81647612 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027480.3 NP_081756.4 16 - 17 1572 71
GenBank mRNA AK043638.1 - 14 - 15 2654 71
AK173316.1 - 14 - 15 2538 71
BC017633.1 - 3 - 4 243 71
BC048236.1 - 15 - 16 1477 71

View Full Product Details