The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023858.3 | NP_076347.3 | 13 - 14 | 1734 | 101 |
NR_024025.1 | 14 - 15 | 1839 | 101 | ||
GenBank mRNA | AK133889.1 | - | 12 - 13 | 1502 | 101 |
AK220369.1 | - | 6 - 7 | 7330 | 101 | |
AY055832.1 | - | 13 - 14 | 2505 | 101 | |
BC027013.1 | - | 4 - 5 | 1311 | 101 | |
BC063050.1 | - | 14 - 15 | 1823 | 101 |