Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tjp3
Entrez Gene ID
27375
Gene Name
tight junction protein 3
Gene Aliases
-
Chromosome Location
Chr.10: 81273188 - 81291581 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282095.1 NP_001269024.1 4 - 5 391 85
NM_001282096.1 NP_001269025.1 4 - 5 454 85
NM_013769.3 NP_038797.2 4 - 5 493 85
GenBank mRNA AF157006.1 - 4 - 5 461 85
AK029083.1 - 4 - 5 440 85
AK134100.1 - 4 - 5 494 85
AK167587.1 - 4 - 5 392 85
BC012518.1 - 4 - 5 458 85
CJ048976.1 - 4 - 5 85

View Full Product Details