Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hmg20b
Entrez Gene ID
15353
Gene Name
high mobility group 20B
Gene Aliases
AW610687, BRAF35, Hmgx2, Hmgxb2, Smarce1r
Chromosome Location
Chr.10: 81346046 - 81350511 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163165.1 NP_001156637.1 5 - 6 647 81
NM_001163166.1 NP_001156638.1 5 - 6 544 81
NM_010440.3 NP_034570.1 4 - 5 731 81
GenBank mRNA AF146224.1 - 4 - 5 714 81
AK153816.1 - 5 - 6 545 81
AK155652.1 - 5 - 6 549 81
AK155890.1 - 4 - 5 607 81
AK186994.1 - 1 - 2 125 81
AK189436.1 - 1 - 2 124 81
AK216551.1 - 1 - 2 124 81
AL355735.1 - 1 - 2 106 81
BC003924.1 - 4 - 5 619 81
BC011334.1 - 5 - 6 580 81
BC023436.1 - 2 - 3 207 81
BC071217.1 - 2 - 3 1608 81

View Full Product Details