The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081375.1 | NP_001074844.1 | 3 - 4 | 304 | 82 |
NM_028219.1 | NP_082495.1 | 4 - 5 | 497 | 82 | |
GenBank mRNA | AK008979.1 | - | 3 - 4 | 305 | 82 |
BC059093.1 | - | 4 - 5 | 476 | 82 |