Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Vps26a
- Entrez Gene ID
-
30930
- Gene Name
- vacuolar protein sorting 26 homolog A (yeast)
- Gene Aliases
-
AA407240, HB58, Vps26
- Chromosome Location
-
Chr.10: 62454843 - 62486805 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC058236.1 |
- |
2 - 3 |
226 |
102 |
View Full Product Details