Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Naalad2
Entrez Gene ID
72560
Gene Name
N-acetylated alpha-linked acidic dipeptidase 2
Gene Aliases
2700022G20Rik, D9Ertd285e, Folh1b, GCP3, GCPIII, NAADALASE2, NAALADASE2
Chromosome Location
Chr.9: 18321954 - 18397538 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028279.3 NP_082555.2 9 - 10 1127 156
GenBank mRNA AK133611.1 - 9 - 10 1237 156
AK135653.1 - 11 - 12 1270 156
AK161457.1 - 12 - 13 1433 156
AY243507.1 - 9 - 10 1127 156
BC108995.1 - 9 - 10 1111 156
BC108996.1 - 9 - 10 1111 156

View Full Product Details