The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023138.4 | NP_075627.2 | 10 - 11 | 1293 | 61 |
GenBank mRNA | AK009392.1 | - | 10 - 11 | 1310 | 61 |
AK011247.1 | - | 10 - 11 | 1287 | 61 | |
AK036822.1 | - | 2 - 3 | 119 | 61 | |
AK080574.1 | - | 10 - 11 | 1294 | 61 | |
AK138130.1 | - | 10 - 11 | 1400 | 61 | |
AK171584.1 | - | 10 - 11 | 1282 | 61 | |
AK172055.1 | - | 10 - 11 | 1291 | 61 | |
AK181352.1 | - | 3 - 4 | 200 | 61 | |
AK186012.1 | - | 3 - 4 | 150 | 61 | |
AK208021.1 | - | 3 - 4 | 150 | 61 | |
BC014830.1 | - | 10 - 11 | 1272 | 61 | |
S68267.1 | - | 10 - 11 | 1214 | 61 |