Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cftr
Entrez Gene ID
12638
Gene Name
cystic fibrosis transmembrane conductance regulator
Gene Aliases
AW495489, Abcc7
Chromosome Location
Chr.6: 18170262 - 18322774 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021050.2 NP_066388.1 11 - 12 1720 132
GenBank mRNA M60493.1 - 11 - 12 1721 132
M69298.1 - 11 - 12 1719 132
M84613.1 - 2 - 3 374 132

View Full Product Details