The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290526.1 | NP_001277455.1 | 4 - 5 | 950 | 75 |
GenBank mRNA | BC158016.1 | - | 4 - 5 | 950 | 75 |
BC172134.1 | - | 4 - 5 | 950 | 75 |