Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Magea2
Entrez Gene ID
17138
Gene Name
melanoma antigen, family A, 2
Gene Aliases
Mage-a2
Chromosome Location
Chr.X: 155027201 - 155033313 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020016.1 NP_064400.1 2 - 3 149 114
GenBank mRNA BC106842.2 - - 49 114

View Full Product Details