The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081403.1 | NP_001074872.1 | 7 - 8 | 1752 | 117 |
GenBank mRNA | AK018108.1 | - | 7 - 8 | 1467 | 117 |
AK173163.1 | - | 7 - 8 | 1466 | 117 |