Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dsc1
Entrez Gene ID
13505
Gene Name
desmocollin 1
Gene Aliases
1110020A10Rik, AI507491, Dsc1a, Dsc1b
Chromosome Location
Chr.18: 20083471 - 20114773 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291804.1 NP_001278733.1 5 - 6 844 62
NM_013504.5 NP_038532.3 5 - 6 844 62
GenBank mRNA AK036893.1 - 5 - 6 730 62
AK079441.1 - 5 - 6 846 62
AK081826.1 - 5 - 6 752 62
BC109161.1 - 5 - 6 681 62
BC109162.1 - 5 - 6 681 62
X97986.1 - 5 - 6 778 62

View Full Product Details