Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ecscr
- Entrez Gene ID
-
68545
- Gene Name
- endothelial cell surface expressed chemotaxis and apoptosis regulator
- Gene Aliases
-
1110006O17Rik, ARIA
- Chromosome Location
-
Chr.18: 35713086 - 35722725 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
EU025067.1 |
- |
1 - 2 |
105 |
66 |
View Full Product Details