The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173383.2 | NP_775559.2 | 2 - 3 | 186 | 101 |
GenBank mRNA | AK141343.1 | - | 2 - 3 | 171 | 101 |
AY321066.1 | - | 2 - 3 | 138 | 101 |