Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Map3k2
- Entrez Gene ID
-
26405
- Gene Name
- mitogen-activated protein kinase kinase kinase 2
- Gene Aliases
-
9630061B06Rik, AI585793, Mekk2, Mekk2b
- Chromosome Location
-
Chr.18: 32163089 - 32236751 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_011946.3 |
NP_036076.2 |
14 - 15 |
1855 |
82 |
View Full Product Details