The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133658.1 | NP_598419.1 | 14 - 15 | 2287 | 74 |
GenBank mRNA | AK151188.1 | - | 14 - 15 | 2292 | 74 |
AK152347.1 | - | 14 - 15 | 2292 | 74 | |
AK152988.1 | - | 14 - 15 | 2299 | 74 | |
AK154745.1 | - | 14 - 15 | 2293 | 74 | |
AK160187.1 | - | 8 - 9 | 1391 | 74 | |
BC016595.1 | - | 14 - 15 | 2258 | 74 | |
BC022956.1 | - | 13 - 14 | 2391 | 74 | |
BC024446.1 | - | 5 - 6 | 633 | 74 | |
BC026575.1 | - | 14 - 15 | 2269 | 74 | |
S71186.1 | - | 14 - 15 | 2287 | 74 |