Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ace2
Entrez Gene ID
70008
Gene Name
angiotensin I converting enzyme (peptidyl-dipeptidase A) 2
Gene Aliases
2010305L05Rik
Chromosome Location
Chr.X: 164139342 - 164188418 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130513.1 NP_001123985.1 3 - 4 677 65
NM_027286.4 NP_081562.2 2 - 3 529 65
GenBank mRNA AB053181.1 - 2 - 3 533 65
AB053182.1 - 2 - 3 533 65
AK141604.1 - 2 - 3 378 65
BC026801.1 - 2 - 3 522 65
EF408739.1 - 3 - 4 638 65
EF408740.1 - 3 - 4 638 65
GQ262785.1 - 2 - 3 347 65

View Full Product Details