The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029872.3 | NP_001025043.3 | 19 - 20 | 2428 | 93 |
GenBank mRNA | AK132915.1 | - | - | 2572 | 93 |
AK156218.1 | - | 19 - 20 | 2429 | 93 |