The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027257.1 | NP_081533.1 | 3 - 4 | 302 | 80 |
GenBank mRNA | AK207620.1 | - | 1 - 2 | 137 | 80 |
AK208217.1 | - | 1 - 2 | 137 | 80 | |
BC026942.1 | - | 3 - 4 | 302 | 80 | |
DQ158906.1 | - | 3 - 4 | 302 | 80 |